Porcine TSH ELISA Kit
Fournisseur: Antibodies.com LLC
ANTIA303664-96EA 685 EUR InStock
ANTIA303664-96
Porcine TSH ELISA Kit
Tests Tests ELISA
Porcine TSH ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of porcine TSH in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready to use ELISA kit
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%
Porcine TSH ELISA kit (A303664) employs the competitive enzyme immunoassay technique for the quantitative measurement of porcine TSH in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with TSH antigen. During the incubation, TSH present in the samples or standards competes with the fixed amount of immobilised TSH for binding sites on the Biotinylated Anti-TSH Antibody. The more TSH present in a sample or standard, the less Biotinylated Anti-TSH Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-TSH Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of TSH present in each sample or standard. The concentration of TSH can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité