Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 610)
Fournisseur: AAT BIOQUEST
50090 50091
AATB50090EA 153 EUR InStock
AATB50090 AATB50091
Anti-IgG Goat Polyclonal Antibody *Cross Adsorbed* (iFluor® 610)
Anticorps
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
  • Affinity purified
  • Easily reconstituted
Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).
Attention: For research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 610
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité