Garder ma session ouverte ?
Expiré dans 
La session est expirée
Votre session a expiré. Pour votre sécurité, VWR vous a déconnecté.
Voulez-vous vous connecter à nouveau?

Human Nesprin1/Syne-1 ELISA Kit

Fournisseur: Antibodies.com LLC
ANTIA313288-96EA 525 EUR InStock
ANTIA313288-96
Human Nesprin1/Syne-1 ELISA Kit
Tests Tests ELISA
Human Nesprin1/Syne-1 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Nesprin1/Syne-1 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 150 to 2400 ng/L
  • Sensitivity: 9,33 ng/L
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Nesprin1/Syne-1 ELISA kit (A313288) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Nesprin1/Syne-1 in serum, plasma or other biological fluids. An antibody specific for Nesprin1/Syne-1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Nesprin1/Syne-1 present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Nesprin1/Syne-1 antibody, which also binds the Nesprin1/Syne-1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Nesprin1/Syne-1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Nesprin1/Syne-1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Nesprin1/Syne-1 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Nesprin1/Syne-1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Commander maintenant
 

COMMANDE

SPÉCIFICATIONS

Saisissez les détails du certificat

Saisissez le numéro de lot :

Choisissez parmi les numéros de lot/lot récents :

Ou
Examinez et comparez les solutions de rechange
VWR a trouvé des produits alternatifs qui peuvent vous sauver jusqu’à . Par unité d'article.  Pour comparer les détails d'un produit, sélectionnez jusqu'à 3 alternatives ci-dessous et cliquez sur Comparer les produits sélectionnés. Pour ajouter des articles à votre panier, entrez une quantité et cliquez sur Ajouter au panier.

Produit original :

Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité