Mouse Anti-Nuclear Antibody ELISA Kit
Fournisseur: Antibodies.com LLC
ANTIA314411-96EA 525 EUR InStock
ANTIA314411-96
Mouse Anti-Nuclear Antibody ELISA Kit
Tests Tests ELISA
Mouse Anti-Nuclear Antibody ELISA kit is a 90 minute Indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of mouse Anti-Nuclear antibody in serum, plasma, and other biological fluids.
  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Mouse anti-Nuclear antibody ELISA kit employs the indirect enzyme-linked immunoassay technique for the qualitative detection of mouse anti-Nuclear antibody in serum, plasma, and other biological fluids. The antigen recognised by the anti-Nuclear antibody has been pre-coated onto a 96-well microtiter plate. Positive controls, negative controls, and samples are added into the wells and incubated. antibodies in the sample bind to the pre-coated antigen. Unbound antibody is removed in a wash step. An HRP-conjugated detection antibody is then added and incubated. Unbound HRP-conjugated detection antibody is removed in another wash step. TMB substrate is then added, and colour develops. The reaction is terminated by the addition of acidic stop solution and the intensity of the yellow colour is measured at O.D. 450 nm in a microplate reader. The O.D. 450 nm of samples is then compared to that of the positive and negative controls to determine the presence or absence of mouse anti-Nuclear antibody.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité