Biopterin ELISA Kit
Fournisseur: Antibodies.com LLC
ANTIA247111-96EA 580 EUR InStock
ANTIA247111-96
Biopterin ELISA Kit
Tests Tests ELISA
Biopterin ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Biopterin in serum, plasma, tissue homogenates, and other biological fluids.
  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Biopterin ELISA kit employs the competitive enzyme immunoassay technique for the quantitative measurement of universal Biopterin in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Biopterin antigen. During the incubation, Biopterin present in the samples or standards competes with the fixed amount of immobilised Biopterin for binding sites on the Biotinylated anti-Biopterin antibody. The more Biopterin present in a sample or standard, the less Biotinylated anti-Biopterin antibody that binds to the plate. Following incubation, unbound Biotinylated anti-Biopterin antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Biopterin present in each sample or standard. The concentration of Biopterin can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité