5-CFDA AM
Fournisseur: AAT BIOQUEST INC
AATB21910EA 127 EUR InStock
AATB21910
5-CFDA AM
5-CFDA AM
5-Carboxyfluorescein diacetate, acetoxymethyl ester (CFDA, AM) is a cell-permeant esterase substrate that can serve as a viability probe that measures both enzymatic activity, which is required to activate its fluorescence, and cell-membrane integrity, which is required for intracellular retention of their fluorescent product.
  • Stable for 12 months
Quality tested in-house
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité