Cell Navigator™ Cell Plasma Membrane Staining Kit, Green
Fournisseur: STEMCELL Technologies
Cell Navigator™
STMC100-0835EA 781 EUR InStock
STMC100-0835
Cell Navigator™ Cell Plasma Membrane Staining Kit, Green
Tests Tests cellulaires
For staining cell plasma membranes in fixed cells - 500 tests.
Cell Navigator™ Cell Plasma Membrane Staining Kits contain a fluorescently labeled dye for staining the plasma membrane in fixed cells. The fluorescently labeled dye stains the plasma membrane without showing cell-type differences as observed with lectins. The dye can be used as a segmentation tool during high-content screening. Cell Navigator™ Cell Plasma Membrane Staining Kit, Deep Red includes Cellpaint™ Deep Red, Live Cell Staining Buffer, and DMSO. Cell Navigator™ Cell Plasma Membrane Staining Kit, Green includes Cellpaint™ Green, Live Cell Staining Buffer, and DMSO.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité