Phalloidin, iFluor™ 647
Fournisseur: STEMCELL Technologies
STMC100-0854EA 324 EUR InStock
STMC100-0854
Phalloidin, iFluor™ 647
Protéines et Peptides
Fluorescent dye conjugates for detection of F-actin in cell culture experiments - 300 tests.
Phalloidin is a heptapeptide derived from the poisonous mushroom Amanita phalloides. Phalloidin binds with a high specificity to F-actin filaments in cells, preventing filament depolymerisation. Fluorescently labeled phalloidin can be used in labelling, identification, and quantitation of F-actin in formaldehyde-fixed and permeabilised cells. Fluorescently labeled phalloidin has lower non-specific binding compared to actin antibodies across most species of plants and animals, which provides low background and high contrast during imaging.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité