LysoBrite™ Lysosome Dye, Red DND-99
Fournisseur: STEMCELL Technologies
LysoBrite™
STMC100-0850EA 324 EUR InStock
STMC100-0850
LysoBrite™ Lysosome Dye, Red DND-99
Systèmes de liaison et de marquage des protéines
Fluorescent dyes used to label and track lysosomes in live cells - 500 tests.
LysoBrite™ dyes are fluorescent dyes that selectively stain lysosomes and other acidic organelles. Due to their hydrophobic nature, LysoBrite™ dyes permeate live cells and selectively accumulate in the lysosomes via the lysosome’s pH gradient. The fluorescence property of these dyes significantly increases within the lysosome’s acidic environment; this fluorescence can be measured using flow cytometry and fluorescence imaging. Due to their selective fluorescence inside lysosomes, LysoBrite™ dyes provide low background and high contrast during imaging. They can be used with proliferating and non-proliferating cells, and are suitable for suspension and adherent cells.
CAS No: 67-68-5
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité