TruCool® Ice Pans with Lids, Multi-Purpose Containers
Fournisseur: BioCision
TruCool®
BCISBCS-117GREA 152 EUR InStock
BCISBCS-117GR BCISBCS-117PK BCISBCS-117PL BCISBCS-118GR BCISBCS-118PL BCISBCS-117B BCISBCS-118B BCISBCS-117OR
TruCool® Ice Pans with Lids, Multi-Purpose Containers
Bacs à glace
Solid, one piece molded Ethylene-vinyl acetate (EVA) foam rectangular ice pans provide unbreakable containment for variety of temperature media.
  • These recyclable ice pans are a non-toxic alternative to conventional ice pans
  • EVA has little or no odor is inert and used in biomedical engineering applications such as drug delivery devices
The multi-purpose containers are highly insulative, stackable, unbreakable, lightweight and leakproof. Containers will not 'sweat' or deform and have a textured finish on the bottom to prevent slipping on the benchtop.
Ideal for containing ice, dry ice, liquid nitrogen, saline solution and acetone. Lid included.
Informations de commande: Available in multiple colors and two sizes, 4 and 9 litre.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité