Industrial Tube Brushes
Fournisseur: JUSTMAN BRUSH CO
715222 715241
JUSB715222EA 17.4 EUR InStock
JUSB715222 JUSB715241
Industrial Tube Brushes
Brosses
Black nylon industrial tube brushes, full range of diameters ranging from 3,2 to 76,2 mm in diameter and up to 1524 mm in length.
Brushes have a wide variety of uses and are typically used for cleaning tubes, industrial tubes, condenser tubes, chiller tubes, valves, small pipes, small tubes and cylinders, bores, tubing lines, water pumps, hydraulic tubes, injection molding, manufacturers, chemical production equipment, oil and gas, farm and farming equipment, automotive cylinders, tubes filled with debris, plumbing tubes and aviation parts and tubes for airplanes of all sizes.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité