Synonyms:
1-Heptyl bromide
A14031.0B
A14031.0I
A14031.30
A14031.0BEA
123
EUR
InStock
A14031.0B
A14031.0I
A14031.30
1-Bromoheptane ≥98%
1-Bromoheptane
Purity: 98+%
CAS No.: 629-04-9
Molecular Formula: CH3(CH2)6Br
Formula Weight: 179.11
Flash Point: 60°(140°F)
Refractive Index: 1.4499
Odour: Recognizable
Melting Point: -58°
Boiling Point: 179-180°
Beilstein Reference: 1697160
UN No.: 1993
MDL No.: MFCD00000273