A11395.0B
A11395.0I
A11395.22
A11395.30
A11395.36
A11395.0BEA
70
EUR
InStock
A11395.0B
A11395.0I
A11395.22
A11395.30
A11395.36
1-Bromo-3-chloropropane ≥99%
1-Bromo-3-chloropropane
Purity: 99%
CAS No.: 109-70-6
Molecular Formula: Cl(CH2)3Br
Formula Weight: 157.44
Flash Point: 140°(284°F)
Refractive Index: 1.4855
Melting Point: -59°
Boiling Point: 142-145°
Beilstein Reference: 605278
UN No.: 2688
MDL No.: MFCD00000998