15294.22EA
118
EUR
InStock
15294.22
15294.36
Cérium (III) acétate 1,5-hydraté ≥99.9% (REO, base d'oxyde de terre rare)
Cérium (III) acétate 1,5-hydraté
With bromide ion, catalyzes the liquid-phase auto-oxidation of cresols.
Cerium(III) acetate sesquihydrate, acts as a source of cerium involved in the preparation of cerium oxide. It finds application in metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. It is utilized in steel production to remove the free sulfur and oxygen. Further, it is an efficient glass polishing agent.