B22884.22
B22884.36
B22884.22EA
55.3
EUR
InStock
B22884.22
B22884.36
1,2-Bis(2-chloroéthoxy)éthane ≥97%
1,2-Bis(2-chloroéthoxy)éthane
Purity: 97%
CAS No.: 112-26-5
Molecular Formula: ClCH2CH2OCH2CH2OCH2CH2Cl
Formula Weight: 187.07
Flash Point: >110°(230°F)
Refractive Index: 1.4610
Melting Point: -32°
Boiling Point: 235°
Beilstein Reference: 506177
UN No.: 2810
MDL No.: MFCD00000976