n-Hexadécane ≥99%
Fournisseur: Thermo Scientific
Danger
A10322.0F A10322.AE A10322.AP
A10322.0FEA 586 EUR InStock
A10322.0F A10322.AE A10322.AP
n-Hexadécane ≥99%
n-Hexadécane
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries.
Purity: 99%
CAS No.: 544-76-3
Molecular Formula: CH3(CH2)14CH3
Formula Weight: 226.45
Flash Point: 135°(275°F)
Physical Form: Liquid
Density: 0.773
Refractive Index: 1.4345
Appearance: Colourless
Melting Point: 18°
Boiling Point: 287°
Solubility: This product is soluble in acetone, alcohol, ether. It is insoluble in water.
Beilstein Reference: 1736592
MDL No.: MFCD00008998
Hexadecane is use in the recovery of valuable amines by means of distillation from aqueous alkanolamine solutions, which are extensively used in the gas-processing industries. *Majid Abedinzadegan Abdi and Axel Meisen. Mutual Solubility of Hexadecane with Diethanolamine and with Bis(hydroxyethyl)piperazine J. Chem. Eng. Data, 1998, 43 (2), pp 138-142
Formule: H₃C(CH₂)₁₄CH₃
MW: 226,45 g/mol
Point d'ébullition: 287 °C
Point de fusion: 18 °C
Densité: 0.773
Point d'éclair: 135 °C (275 °F)
Température de stockage: Température ambiante
Numéro MDL: MFCD00008998
Numéro CAS: 544-76-3
EINECS: 208-878-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90d3084bdd908b305ecf9b3c30e03cd98a407d94&vid=e5a15f4f6c36024a34521e08083957252ff78457&prid=web&clid=SAPDAM
Commander maintenant
COMMANDE
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité