A15118.14EA
164
EUR
InStock
A15118.14
A15118.22
A15118.06
2,2,6,6-Tétraméthyl-3,5-heptanedione ≥98%
2,2,6,6-Tétraméthyl-3,5-heptanedione
Purity: 98%
CAS No.: 1118-71-4
Molecular Formula: C11H20O2
Formula Weight: 184.28
Flash Point: 67°(152°F)
Refractive Index: 1.4590
Boiling Point: 72-73°/6mm
Beilstein Reference: 1447269
MDL No.: MFCD00008848