L02260.14EA
48.4
EUR
InStock
L02260.14
Isopentylamine 99%
Isopentylamine
Purity: 99%
CAS No.: 107-85-7
Molecular Formula: (CH3)2CHCH2CH2NH2
Formula Weight: 87.17
Flash Point: -1°(30°F)
Refractive Index: 1.4080
Odour: Amine-like
pH: 12.1
Melting Point: -60°
Boiling Point: 95-97°
Beilstein Reference: 1209230
Merck Reference: 14,5112
UN No.: 1106
MDL No.: MFCD00008203