B23778.06
B23778.14
B23778.06EA
56.2
EUR
InStock
B23778.06
B23778.14
1,2,4-Tribromobenzène 95%
1,2,4-Tribromobenzène
Purity: 95%
CAS No.: 615-54-3
Molecular Formula: C6H3Br3
Formula Weight: 314.80
Flash Point: >110°(230°F)
Refractive Index: >110^o(230^oF)
Melting Point: 41-43°
Boiling Point: 274-276°
Beilstein Reference: 1934823
MDL No.: MFCD00000062