A14080.22EA
152
EUR
InStock
A14080.22
A14080.14
2,3-Diméthoxybenzaldéhyde ≥98%
2,3-Diméthoxybenzaldéhyde
Purity: 98+%
CAS No.: 86-51-1
Molecular Formula: C9H10O3
Formula Weight: 166.18
Flash Point: >110°(230°F)
Melting Point: 51-52°
Boiling Point: 178°/20mm
Beilstein Reference: 908264
MDL No.: MFCD00003309