A15109.22EA
678
EUR
InStock
A15109.22
A15109.14
A15109.06
3,5-Diméthoxybenzaldéhyde ≥98%
3,5-Diméthoxybenzaldéhyde
Purity: 98%
CAS No.: 7311-34-4
Molecular Formula: C9H10O3
Formula Weight: 166.18
Flash Point: >110°(230°F)
Melting Point: 44-48°
Boiling Point: 115-118°/1mm
Beilstein Reference: 2042374
MDL No.: MFCD00003366