A13688.0B
A13688.30
A13688.36
A13688.0BEA
148
EUR
InStock
A13688.0B
A13688.30
A13688.36
1,3-Dichlorobenzène ≥98%
1,3-Dichlorobenzène
Purity: 98%
CAS No.: 541-73-1
Molecular Formula: C6H4Cl2
Formula Weight: 147.00
Flash Point: 63°(145°F)
Refractive Index: 1.5460
Melting Point: -25°
Boiling Point: 173°
Beilstein Reference: 956618
Merck Reference: 14,3055
UN No.: 3082
MDL No.: MFCD00000573