B21511.14EA
149
EUR
InStock
B21511.14
1,2,3-Trichloro-5-nitrobenzène ≥97%
1,2,3-Trichloro-5-nitrobenzène
Purity: 97%
CAS No.: 20098-48-0
Molecular Formula: C6H2Cl3NO2
Formula Weight: 226.45
Melting Point: 68-71°
Beilstein Reference: 2505480
UN No.: 2811
MDL No.: MFCD00061096