A12144.14EA
37.3
EUR
InStock
A12144.14
A12144.36
A12144.22
4-Iodoanisole ≥98%
4-Iodoanisole
Purity: 98+%
CAS No.: 696-62-8
Molecular Formula: C7H7IO
Formula Weight: 234.04
Flash Point: >110°(230°F)
Melting Point: 48-52°
Boiling Point: 240°
Beilstein Reference: 1906692
MDL No.: MFCD00001056