A15312.09EA
40.4
EUR
InStock
A15312.09
3-Nitrophénol ≥98%
3-Nitrophénol
Purity: 98+%
CAS No.: 554-84-7
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Flash Point: >100°(212°F)
Melting Point: 93-98°
Boiling Point: 194°/70mm
Beilstein Reference: 1907946
Merck Reference: 14,6619
UN No.: 1663
MDL No.: MFCD00007240