Synonyms:
2-Bromobenzyl bromide
A11285.09
A11285.18
A11285.30
A11285.09EA
56.6
EUR
InStock
A11285.09
A11285.18
A11285.30
α,2-Dibromotoluène ≥98%
α,2-Dibromotoluène
Purity: 98%
CAS No.: 3433-80-5
Molecular Formula: C7H6Br2
Formula Weight: 249.94
Flash Point: >110°(230°F)
Refractive Index: 1.6190
Melting Point: 29-32°
Boiling Point: 127-129°/19mm
Beilstein Reference: 971015
UN No.: 1759
MDL No.: MFCD00000173