L20093.22EA
268
EUR
InStock
L20093.22
2-Fluoro-6-nitrotoluène ≥98%
2-Fluoro-6-nitrotoluène
Purity: 98%
CAS No.: 769-10-8
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Flash Point: 88°(190°F)
Refractive Index: 1.5240
Melting Point: 6-8°
Boiling Point: 208-210°
Beilstein Reference: 2361978
UN No.: 2810
MDL No.: MFCD00007197