A12941.14EA
135
EUR
InStock
A12941.14
A12941.06
(2,2-Diméthylpropyl)benzène ≥97%
(2,2-Diméthylpropyl)benzène
Purity: 97+%
CAS No.: 1007-26-7
Molecular Formula: C11H16
Formula Weight: 148.25
Flash Point: 50°(122°F)
Refractive Index: 1.4880
Boiling Point: 184-185°
Beilstein Reference: 2037544
UN No.: 3295
MDL No.: MFCD00038279