A13109.06EA
86
EUR
InStock
A13109.06
A13109.14
3-Méthylquinoléine ≥98%
3-Méthylquinoléine
Purity: 98+%
CAS No.: 612-58-8
Molecular Formula: C10H9N
Formula Weight: 143.19
Flash Point: >110°(230°F)
Refractive Index: 1.6150
Melting Point: 15°
Boiling Point: 252-253°
Beilstein Reference: 110325
MDL No.: MFCD00014661