L09469.06EA
64.8
EUR
InStock
L09469.06
1,1,2-Trichloro-2,3,3-trifluorocyclobutane ≥98%
1,1,2-Trichloro-2,3,3-trifluorocyclobutane
Purity: 98%
CAS No.: 697-17-6
Molecular Formula: C4H2Cl3F3
Formula Weight: 213.41
Flash Point: None
Refractive Index: 1.4160
Boiling Point: 119-120°
Beilstein Reference: 1905850
MDL No.: MFCD00039439