L13015.06EA
210
EUR
InStock
L13015.06
Fluorocyclohexane ≥97%
Fluorocyclohexane
Purity: 97%
CAS No.: 372-46-3
Molecular Formula: C6H11F
Formula Weight: 102.15
Flash Point: 5°(41°F)
Refractive Index: 1.4160
Melting Point: 13°
Boiling Point: 103-105°
Beilstein Reference: 1900795
UN No.: 1993
MDL No.: MFCD00021285