Synonyms:
Octamethylene
L15000.AD
L15000.AK
L15000.ADEA
50
EUR
InStock
L15000.AD
L15000.AK
Cyclooctane ≥99%
Cyclooctane
Purity: 99+%
CAS No.: 292-64-8
Molecular Formula: C8H16
Formula Weight: 112.22
Flash Point: 28°(82°F)
Refractive Index: 1.4580
Melting Point: 12-14°
Boiling Point: 150-152°
Beilstein Reference: 1900349
UN No.: 3295
MDL No.: MFCD00004162