12585.22
12585.36
12585.22EA
75.3
EUR
InStock
12585.22
12585.36
Sodium sélénite, anhydre ≥99% (base métaux) typ. ≥ 99.75%
Sodium sélénite
Purity: 99% min
CAS No.: 10102-18-8
Molecular Formula: Na2O3Se
Formula Weight: 172.94
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
pH: 9
Melting Point: 350°
Solubility: Freely soluble in water. Insoluble in alcohol
Merck Reference: 14,8673
UN No.: 2630
MDL No.: MFCD00003489
In glass manufacturing