Synonyms:
1,5,5-Trimethylimidazolidine-2,4-dione
B24188.06
B24188.14
B24188.22
B24188.06EA
52.4
EUR
InStock
B24188.06
B24188.14
B24188.22
1,5,5-Triméthylhydantoïne ≥98%
1,5,5-Triméthylhydantoïne
Purity: 98%
CAS No.: 6851-81-6
Molecular Formula: C6H10N2O2
Formula Weight: 142.16
Melting Point: 161-164°
MDL No.: MFCD00040439