L13454.03
L13454.06
L13454.14
L13454.03EA
37.7
EUR
InStock
L13454.03
L13454.06
L13454.14
4-Méthyl-3-nitrobenzaldéhyde ≥98%
4-Méthyl-3-nitrobenzaldéhyde
Purity: 98%
CAS No.: 31680-07-6
Molecular Formula: C8H7NO3
Formula Weight: 165.15
Flash Point: 110°(230°F)
Melting Point: 44-48°
Beilstein Reference: 1637706
MDL No.: MFCD00017011