Cyclosporin A, analytical standard, VETRANAL™, Supelco®
Fournisseur: Merck
VETRANAL™
32425-100MGEA 267 EUR InStock
32425-100MG
Cyclosporin A, analytical standard, VETRANAL™, Supelco®
Étalons Étalons organiques
Cyclosporin A (CsA) is a fungal metabolite, a cyclic undecapeptide. It has highly specific immunosuppressive effects with low myelotoxicity, thereby making it a valuable drug in human organ transplantation. Its immunosuppressive activity is based upon its ability to inhibit early steps of T-lymphocyte activation such as lymphokine gene transcription.
A fungal metabolite possessing potent immunosuppressive properties. It inhibits the T-cell receptor signal transduction pathway via the formation of cyclosporin A−cyclophilin complex that inhibits calcineurin (protein phosphatase 2B). Inhibits nitric oxide synthesis induced by interleukin 1α, lipopolysaccharides and TNFα. Can block cytochrome c release from mitochondria.
Attention: Signal word: Danger
GHS Symbols: GHS08, GHS07
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité