Synonyms:
2-n-Propylresorcinol
H27024.06
H27024.03
H27024.06EA
268
EUR
InStock
H27024.06
H27024.03
2-Propylrésorcinol ≥98%
2-Propylrésorcinol
Purity: 98%
CAS No.: 13331-19-6
Molecular Formula: C9H12O2
Formula Weight: 152.19
Flash Point: 135°(275°F)
Odour: Phenol-like
Boiling Point: 280°
MDL No.: MFCD00100568