L14221.AC
L14221.AE
L14221.ACEA
29.7
EUR
InStock
L14221.AC
L14221.AE
Cyclohexyl(éthyl)amine 97%
Cyclohexyl(éthyl)amine
Purity: 97%
CAS No.: 5459-93-8
Molecular Formula: C8H17N
Formula Weight: 127.23
Flash Point: 45°(113°F)
Refractive Index: 1.4520
Odour: Amine-like
Boiling Point: 163-166°
Beilstein Reference: 2070280
UN No.: 2734
MDL No.: MFCD00003834