Synonyms:
6-Nitro-4H-chromen-4-one
H27167.14
H27167.06
H27167.03
H27167.14EA
246
EUR
InStock
H27167.14
H27167.06
H27167.03
6-Nitrochromone ≥97%
6-Nitrochromone
Purity: 97%
CAS No.: 51484-05-0
Molecular Formula: C9H5NO4
Formula Weight: 191.15
Melting Point: 172-175°
MDL No.: MFCD02954226