Improve Your Sample Preparation
Effective Tissue Disruption & Cell Lysis
In most Life Science workflows, one of the first steps is to disrupt your sample with a tissue homogenizer before the extraction of molecules of interest.
Using highly sophisticated bead-beating technology the Precellys® and Minilys® bench top homogenisers offer great benefits!
  • Highest yields: Efficient lysis and minimal sample loss - cooling option to protect sensitive target molecules
  • Time saving: Homogenisation takes seconds not minutes - process up to 24 samples in parallel
  • Reproducibility: Programmable and storable settings - no user fatigue or variance between individual samples
  • Contamination-free: Closed tube approach - no contact between sample and environment
  • Widest application range: Flexible grinding parameters - huge choice of optimised lysing kits in different tube sizes
Discover numerous established Protocols
Precellys Evolution – SUPER homogenizer
Precellys Evolution in Spotlight
The Precellys® Evolution offers optimal robustness, safety, efficiency and flexibility. Especially designed for Bio-safety class L3 laboratories, the Precellys® Evolution is based on 10 years of sample preparation expertise.
    Precellys in action!
    Product Selection
    Extract & Purify
    Also interested in solutions for nucleic acid purification? RNA, genomic DNA or plasmids from various sources? Clean-up of PCR or labelling reactions? Manual or automated sample processing?
      Check out what VWR can offer you