Easy, automated cellular imaging and analysis
|
Applications
- Cell counting and cell viability
- Neurite tracing
- Apoptosis
- Phagocytosis, endocytosis and internalization
- Viral infectivity assays
- Autophagy detection
- DNA damage assays
Product Details |
ImageXpress® Nano (2-60x objective) The ImageXpress® Nano Automated Imaging System enables researchers to get better data faster and improve collaborations with peers. An easy to use, browser based interface, compatible with touch screens and tablets, allows on the fly image analysis and access to data anywhere, anytime. This cost-effective automated microscopy system allows you to scale up your experiments to multi-slide or microplate-level evaluations in a system that can be configured to match your needs and to configure your system with your preferred filters and objectives. Additional options include transmitted light capability for imaging of non-labeled samples and live cell assays provided through our environmental control feature (temperature, humidity, CO2). Request a Demo |