Frac30 fraction collector
Frac30 fraction collector
Frac30, a round fraction collector, is controlled via the ÄKTA start touch screen instrument display or using UNICORN start software on a computer.
Frac30 allows you to collect up to 30 fractions and supports four tube sizes, ranging from 1.5 mL to 15 mL. Fractions can be collected in volumes ranging from 0.5 mL to 15 mL. The entire fraction collector rack can be transferred as required. 
Depletion of albumin from serum samples using ÄKTA start
Purification of GST-tagged protein using ÄKTA start
Purification of an N-terminal histidine-tagged protein using ÄKTA start
Purification of antibodies using ÄKTA start and HiTrap Protein G HP column
Product Information
29-0187-51 - UNICORN™ start software
29-0230-51 - Frac30 fraction collector
29-0220-94 - ÄKTA™ start, main instrument