Protexidase™, peroxidase protection buffer, Immuno™, MP Biomedicals
Fournisseur: MP Biomedicals
08980631
ICNA08980631EA 673 EUR InStock
ICNA08980631
Protexidase™, peroxidase protection buffer, Immuno™, MP Biomedicals
Buffers Tampons biologiques
A peroxidase protective buffer which stabilises HRP conjugates. It has 500 ml of PBS, pH 7,2, containing 0,5% BSA, antimicrobial preservatives and proprietary stabilising compounds.
It is compatible with all common blocking agents including Bovine Serum Albumin, skim milk powder and Polysorbate 20. Protexidase provides stability of the HRP enzyme and it also inhibits microbial growth in dilute conjugates.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité