Pierce™ EndoProteinase Lys-C Protease, MS Grade
Fournisseur: Thermo Fisher Scientific
90051
PIER90051EA 444 EUR InStock
PIER90051
Pierce™ EndoProteinase Lys-C Protease, MS Grade
Protéines extraction Réactifs pour l'extraction des protéines
Endoproteinase Lys-C protease is a highly purified native endoproteinase validated for maximum activity and stability in proteomic applications.
  • Can be used in tandem with trypsin to decrease tryptic missed cleavages thus enhancing digestion
  • Better protein characterisation results from overlapping peptides with complementary chromatographic, ionisation and fragmentation properties thereby increasing sequence coverage
  • Effective enzyme activity under highly denaturing conditions
  • C-terminal lysine cleavage specificity of 90% for a complex protein sample
  • Stable since it is provided in a lyophilised format
Applications include improved sequence coverage of protein digests, de novo sequencing, epigenetic studies and in-gel and in-solution digestion of proteins.
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité