Anti-DPF3 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
25-194
PRSI25-194EA 660 EUR InStock
PRSI25-194
Anti-DPF3 Rabbit Polyclonal Antibody
Anticorps
DPF3 is a muscle-specific component of the BAF complex, a multiprotein complex involved in transcriptional activation and repression of select genes by chromatin remodeling (alteration of DNA-nucleosome topology). DPF3 specifically binds acetylated lysines on histone 3 and 4 (H3K14ac, H3K9ac, H4K5ac, H4K8ac, H4K12ac, H4K16ac). In the complex, it acts as a tissue-specific anchor between histone acetylations and methylations and chromatin remodeling. It thereby probably plays an essential role in heart and skeletal muscle development.
DPF3 antibody can be used for detection of DPF3 by ELISAMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité