Anti-PCDHGA4 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
26-396
PRSI26-396EA 660 EUR InStock
PRSI26-396
Anti-PCDHGA4 Rabbit Polyclonal Antibody
Anticorps
PCDHGA4 is a single-pass type I membrane protein. It contains 6 cadherin domains. PCDHGA4 is a potential calcium-dependent cell-adhesion protein. It may be involved in the establishment and maintenance of specific neuronal connections in the brain.This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité