Anti-FUBP3 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
26-610
PRSI26-610EA 660 EUR InStock
PRSI26-610
Anti-FUBP3 Rabbit Polyclonal Antibody
Anticorps
FUBP3 may interact with single-stranded DNA from the far-upstream element (FUSE). FUBP3 may activate gene expression.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FUBP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité