Anti-LYPLA2 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
27-202
PRSI27-202EA 660 EUR InStock
PRSI27-202
Anti-LYPLA2 Rabbit Polyclonal Antibody
Anticorps
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids.Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. There are alternatively spliced transcript variants described for this gene but the full length nature is not known yet.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. AvoidMore Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité