Anti-POU1F1 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
27-336
PRSI27-336EA 601 EUR InStock
PRSI27-336
Anti-POU1F1 Rabbit Polyclonal Antibody
Anticorps
PIT1 is a pituitary-specific transcription factor responsible for pituitary development and hormone expression in mammals and is a member of the POU family of transcription factors that regulate mammalian development. The POU family is so named because the first 3 members identified were PIT1 and OCT1 of mammals, and Unc-86 of C. elegans. PIT1 contains 2 protein domains, termed POU-specific and POU-homeo, which are both necessary for high affinity DNA binding on genes encoding growth hormone and prolactin. PIT1 is also important for regulation of the genes encoding prolactin and thyroid-More Product Information
Collapse All
Taille Cdt Numéro de catalogue VWR Quantité
100 µg 100 µG PRSI27-336
claquement
Veuillez sélectionner l'article
Entrer les détails
SPÉCIFICATIONS
Documentation
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité