Anti-KIFC2 Rabbit Polyclonal Antibody
Fournisseur: ProSci Inc.
27-982
PRSI27-982EA 601 EUR InStock
PRSI27-982
Anti-KIFC2 Rabbit Polyclonal Antibody
Anticorps
Members of the kinesin superfamily of microtubule-associated proteins are involved in a variety of intracellular processes including cell division and organelle transport. KIFC2 encodes a 792 amino acid protein, which contains the conserved motor domain at the C-terminal end of the protein and is most similar to members of the KAR3 family involved in cell division.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below.More Product Information
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité